Fisher Chemical Sodium Acetate Anhydrous, 1 kg

Representative image only.

Fisher Chemical Sodium Acetate Anhydrous, 1 kg

Fisher Chemical | Mfr: BP3331| Item: UX-88064-72

Quality and consistency your lab can count on

$356.25USD / Each

$356.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeNone
  • Empirical FormulaC2H3O2Na
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)1000
  • Formula Weight82.03
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number127-09-3
  • Melting Point C (° C)324
  • DescriptionSodium Acetate Anhydrous, 1 kg
Documents & Certificates
Reviews
Q&A