Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 500 g

Representative image only.

Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 500 g

Fisher Chemical | Mfr: S318500| Item: UX-88024-06

Quality and consistency your lab can count on

$178.00USD / Each

$178.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥97.0%
  • Sodium Carbonate — ≤1.0%
  • Chloride — ≤0.005%
  • Nitrogen Compounds (as N) — ≤0.001%
  • Phosphate — ≤0.001%
  • Sulfate — ≤0.003%
  • Ammonium Hydroxide Precipitate — ≤0.020%
  • Heavy Metals (as Ag) — ≤0.002%
  • Iron — ≤0.001%
  • Nickel — ≤0.001%
  • Potassium — ≤0.02%
  • Mercury — ≤0.1ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

CorrosiveStorage Code White
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaNaOH
  • ClassReagent Acids and Bases
  • ChemistryAnalytical Reagents
  • SynonymsLye, Caustic Soda
  • FormSolid
  • Weight (g)500
  • Formula Weight40.00
  • PackagingPoly Bottle
  • DOT InformationSodium hydroxide, solid; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2
  • UN NumberUN1823
  • Boiling Point C (° C)1390
  • Melting Point C (° C)318
  • DescriptionSodium Hydroxide, Certified ACS Grade, 500 g
Documents & Certificates
Reviews
Q&A