Fisher Chemical Sodium Hydroxide Solution, 0.1N (N/10), Certified Grade, 1 L

Representative image only.

Fisher Chemical Sodium Hydroxide Solution, 0.1N (N/10), Certified Grade, 1 L

Fisher Chemical | Mfr: SS2761| Item: UX-88024-48

Quality and consistency your lab can count on

$94.20USD / Each

$94.20USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Normality — 0.0995-0.1005
  • Color (APHA) — ≤10

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

CorrosiveStorage Code White
Specifications
  • Normality0.1
  • Purity GradeCertified
  • Concentration0.1N
  • Empirical FormulaNaOH
  • MatrixWater
  • ClassReagent Acids and Bases
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda
  • Volume (mL)1000
  • Formula Weight40
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2
  • Boiling Point C (° C)100
  • Melting Point C (° C)0
  • DescriptionSodium Hydroxide Solution, 0.1N (N/10), Certified Grade, 1 L
Documents & Certificates
Reviews
Q&A