Fisher Chemical Tetrahydrofuran, Certified Grade, 1 L

Representative image only.

Fisher Chemical Tetrahydrofuran, Certified Grade, 1 L

Fisher Chemical | Mfr: T3971| Item: UX-88040-20

Quality and consistency your lab can count on

$445.25USD / Each

$445.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤10
  • Boiling Range — 65°-67°C
  • Specific Gravity at 25°C — 0.884-0.887
  • Peroxides (as THF Peroxide) — ≤0.015%
  • Water — ≤0.02%
  • Inhibitor (BHT) — 75-400ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableIrritantStorage Code Red
Specifications
  • Purity GradeCertified
  • Empirical FormulaCH2(CH2)2CH2O
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsTHF
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight72.11
  • PackagingGlass Bottle
  • DOT InformationTetrahydrofuran; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolHighly Flammable, Irritant
  • UN NumberUN2056
  • CAS Number109-99-9
  • Boiling Point C (° C)66
  • Melting Point C (° C)-108.4
  • DescriptionTetrahydrofuran, Certified Grade, 1 L
Documents & Certificates
Reviews
Q&A