Fisher Chemical Tetrahydrofuran, OPTIMA™ Grade, 1 L

Representative image only.

Fisher Chemical Tetrahydrofuran, OPTIMA™ Grade, 1 L

Fisher Chemical | Mfr: T4271| Item: UX-88040-04

Quality and consistency your lab can count on

$305.75USD / Each

$305.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Color (APHA) — ≤10
  • Refractive Index at 25°C — 1.4040-1.4070
  • Residue after Evaporation by Steam Bath — ≤5ppm
  • Water — ≤0.02%
  • Fluorescence Background (as Quinine Sulfate) — ≤1ppb
  • Peroxides (as THF Peroxide) — ≤0.015%
  • Optical Absorbance at 210nm — ≤1.00
  • at 215nm — ≤0.60
  • at 230nm — ≤0.30
  • at 254nm — ≤0.10

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableIrritantStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • Empirical FormulaCH2(CH2)2CH2O
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsTHF
  • Volume (mL)1000
  • Formula Weight72.11
  • PackagingAmber Glass Bottle
  • DOT InformationTetrahydrofuran; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • CAS Number109-99-9
  • UN NumberUN2056
  • Boiling Point C (° C)66
  • Melting Point C (° C)-108.4
  • DescriptionTetrahydrofuran, OPTIMA™ Grade, 1 L
Documents & Certificates
Reviews
Q&A