Fisher Chemical Tris Hydrochloride Solution, 1M, pH 7, Molecular Biology Grade, 500 mL

Representative image only.

Fisher Chemical Tris Hydrochloride Solution, 1M, pH 7, Molecular Biology Grade, 500 mL

Fisher Chemical | Mfr: BP1756100| Item: UX-88058-08

Quality and consistency your lab can count on

$257.00USD / Each

$257.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Concentration1M
  • Purity GradeMolecular Biology
  • ChemistrySolutions
  • FormLiquid
  • SynonymsTriethanolamine, Tris
  • Volume (mL)100
  • Formula Weight121.1
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGrey
  • CAS Number77-86-1 (Tris) 1185-53-1 (Tris HCl)
  • DescriptionTris Hydrochloride Solution, 1M, pH 7, Molecular Biology Grade, 500 mL
Documents & Certificates
Reviews
Q&A