Fluid D, USP, prepared; 1 liter bottle, 10/pk

Representative image only.

Fluid D, USP, prepared; 1 liter bottle, 10/pk

| Item: UX-14204-62

Save time and money with ready-made bottles

$366.50USD / Pkg of 10

$366.50USD / Pkg of 10
Product Description

Key Features

  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • A rinsing and diluting fluid for use in USP testing protocols
  • pH 7.1
  • Contains peptic digest of animal tissue and polysorbate 80
  • 6 month warranty

More About this Item

Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

6 Month Warranty
Specifications
  • Media Name (No.)Fluid D
  • Capacity (mL)1000
  • Capacity (L)1
  • DescriptionFluid D, USP, prepared; 1 liter bottle, 10/pk
  • Warranty6 month
Reviews
Q&A