Fluke Calibration 2373-LPRT Adapter, Lemo to Mini Grabbers (4-Wire)
Representative image only.
Fluke Calibration 2373-LPRT Adapter, Lemo to Mini Grabbers (4-Wire)
Fluke Calibration | Mfr: 2373-LPRT| Item: UX-93202-57
Use to connect 1523 or 1524 reference thermometer to the leads of the RTD you want to measure
$373.50USD / Each
$415.00USD / Each
$373.50USD / Each
$415.00USD / Each
Product Description
Key Features
  • Adapters provide an easy way to connect PRTs and RTD leads
  • The 4-wire mini-grabbers make it easy to connect and disconnect
  • 1 year warranty
More About this Item
Keep your thermometers running at their best with these accessories.
1 Year Warranty
Specifications
  • Compatible WithFluke Calibration 1523 and 1524 thermometers
  • ConnectionsLemo to mini grabbers (4-wire)
  • DescriptionAdapter, Lemo to Mini Grabbers (4-Wire)