Fluke SBP810 Replacement Lithium-Ion Battery Pk, 14.8 V, 2.55 Ah

Representative image only.

Fluke SBP810 Replacement Lithium-Ion Battery Pk, 14.8 V, 2.55 Ah

Fluke | Mfr: SBP810| Item: UX-65700-27

$394.06USD / Each
$437.85USD / Each

$394.06USD / Each
$437.85USD / Each
Product Description

More About this Item

Keep your vibration tester running smoothly with these power sources.

Specifications
  • DescriptionReplacement Lithium-Ion Battery Pk, 14.8 V, 2.55 Ah
Reviews
Q&A