Fluke TP220 Industrial Test Probes
Representative image only.
Fluke TP220 Industrial Test Probes
Fluke | Mfr: TP220| Item: UX-26002-97
High performance at an economical price
$31.50USD / Set of 2
$35.00USD / Set of 2
$31.50USD / Set of 2
$35.00USD / Set of 2
Buy It Together
Current Item
Fluke TP220 Industrial Test Probes
Fluke TP220 Industrial Test Probes
2600297
$31.50USD / Set of 2
$35.00USD / Set of 2
+
Fluke TL224 Flexible Silicone Test Lead Set
2600225
$42.30USD / Set of 2
$47.00USD / Set of 2
=
Total Price 73.80 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Very sharp, extra-long 0.750" (19 mm) stainless steel tip. For reaching deep test points or recessed components; will pierce insulation.
One red, one black. 1000 V 10 A maximum.
Available for sale in the United States and Canada only
1 Year Warranty
Specifications
  • DescriptionIndustrial Test Probes