Graphic Controls 00006932 Circular Chart, 7 day, 12" dia, 0-100 scale (x2)
Representative image only.
Graphic Controls 00006932 Circular Chart, 7 day, 12" dia, 0-100 scale (x2)
Graphic Controls | Mfr: 00006932| Item: UX-49000-45
Get exacting performance that meets American Gas Association (AGA) standards
$61.39USD / Box of 100
$87.70USD / Box of 100
$61.39USD / Box of 100
$87.70USD / Box of 100
Product Description
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Dual scales read 0 to 100
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.
1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)12
  • DescriptionCircular Chart, 7 day, 12" dia, 0-100 scale (x2)