Guardian Equipment G1100 Faucet-Mount Personal Eyewash
Representative image only.
Guardian Equipment G1100 Faucet-Mount Personal Eyewash
Guardian Equipment | Mfr: G1100| Item: UX-06767-42
Installs easily on standard faucet; water pressure keeps stream flowing
$122.55USD / Each
$171.50USD / Each
$122.55USD / Each
$171.50USD / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Adjustable outlets are 3" (7.6 cm) apart and angled forward. Chrome-plated brass construction.
Compliance: ANSI Z358.1-2004 as personal eyewash only.
Two year warranty
Specifications
  • Eyewash Fixture MaterialChrome-plated brass
  • Supply Pipe MaterialChrome-plated brass
  • MountFaucet Mount
  • Inlet Connection55/64"-27 (F) for faucet
  • DescriptionFaucet-Mount Personal Eyewash