Hach 1454201 Free Chlorine Test Kit , Range 0.02 to 0.68 and 0 to 3.4 ppm
Representative image only.
Hach 1454201 Free Chlorine Test Kit , Range 0.02 to 0.68 and 0 to 3.4 ppm
Hach | Mfr: 1454201| Item: UX-05545-68
Color wheel method provides accurate visual comparisons
$152.00USD / Each
$152.00USD / Each
Buy It Together
Current Item
Hach 1454201 Free Chlorine Test Kit , Range 0.02 to 0.68 and 0 to 3.4 ppm
Hach 1454201 Free Chlorine Test Kit , Range 0.02 to 0.68 and 0 to 3.4 ppm
0554568
$152.00USD / Each
+ + =
Total Price 930.10 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, free
  • Measurement Points0 to 3.4 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit100
  • DescriptionFree Chlorine Test Kit , Range 0.02 to 0.68 and 0 to 3.4 ppm