Hach 223101 Chlorine Free and Total Color Disc Test Kit
Representative image only.
Hach 223101 Chlorine Free and Total Color Disc Test Kit
Hach | Mfr: 223101| Item: UX-05545-72
Color wheel method provides accurate visual comparisons
$112.00USD / Each
$112.00USD / Each
Buy It Together
Current Item
Hach 223101 Chlorine Free and Total Color Disc Test Kit
Hach 223101 Chlorine Free and Total Color Disc Test Kit
0554572
$112.00USD / Each
+ + =
Total Price 890.10 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, free and total
  • Measurement Points0 to 3.4 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit100
  • DescriptionChlorine Free and Total Color Disc Test Kit