Hardman Double Bubble® Urethane Pouches, Flexible High Peel Strength, 3.5g; 10/Pk
Representative image only.
Hardman Double Bubble® Urethane Pouches, Flexible High Peel Strength, 3.5g; 10/Pk
Hardman | Mfr: 4022-BG10| Item: UX-08778-22
Complete selection of epoxies and urethane for all your maintenance needs
$45.40USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$45.40 USD / Pkg of 10
  • 10+$40.86 USD / Pkg of 10
$45.40USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$45.40 USD / Pkg of 10
  • 10+$40.86 USD / Pkg of 10
Product Description
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • A good combination of high peel and lap shear strengths
  • It is semi-flexible and will bond to rubber, leather, china and many plastics
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.
Specifications
  • Curing time (days)1
  • Curing time (mins)1440
  • Curing time (hrs)24
  • Set time (mins)8
  • Min operating temperature (° F)-40
  • Max operating temperature (° F)175
  • Number of Pieces per Set10
  • DescriptionUrethane Pouches, Flexible High Peel Strength, 3.5g; 10/Pk