Product Type
In Stock
Length (in)
Industry
Regulatory Compliance
US Origin
Megohmmeters
The product searched is discontinued, please browse below for replacements.
Megohmmeters
Megohmmeters help you ensure the safety of electrical installations and equipment. A portable megohmmeter is a rugged unit that can perform voltage, insulation, and capacitance measurements quickly and accurately. These units are designed for conducting high resistance or absorption tests, time resistance measurements, DAR and PI test, and are ideal for use on wiring installations, transformers, motors, generators, and more. Many of our megohmmeters are designed with built-in safety features like automatic test inhibition capability that will not allow test voltages to be generated if a live sample is detected.