Honeywell S2970HS Uvex Hypershock Safety Glasses, Clear Frame, Clear, HydroShield Antifog Lens; 10/BX
Representative image only.
Honeywell S2970HS Uvex Hypershock Safety Glasses, Clear Frame, Clear, HydroShield Antifog Lens; 10/BX
Honeywell | Mfr: S2970HS| Item: EW-86497-98
High-performance wraparound eyewear with a sporty style
$244.50USD / Box of 10
$244.50USD / Box of 10
Product Description
Key Features
  • Protects against impact from sun, wind and dust, as well as airborne debris HydroShield long-life antifog coating to help keep lenses clear Soft molded inserts and nosepiece provide a comfortable, secure fit
  • Clear Frame
More About this Item
Sharp and modern looking, these clear safety glasses have superior side protection and peripheral vision. Hypershock glasses are 100-percent dielectric so, you can wear them in environments where accidental exposure to an electrical charge is possible. Polycarbonate lens features antiscratch and HydroShield antifog hard coatings.
Compliance: ANSI Z87.1-2015, CSA Z94.3
Specifications
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1-2015, CSA Z94.3
  • DescriptionSafety Glasses, Clear Frame, Clear, Hydroshield Antifog Lens; 10/BX