ICI Quick Ship Bench Fume Hood Work Surface, without Cup Sink Cutout, 72" Wide

Representative image only.
ICI Quick Ship Mfr # CPWL-72B – Item # EW-78981-07
Enhance your fume hood performance
  • Dished to retain spills,
  • Ideal for use with 72" wide bench fume hood

PRODUCT OPTIONS

$3,928.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithICI Quick Ship ducted fume hoods
  • Width (in)72
  • Width (cm)182.9
  • DescriptionICI Quick Ship Bench Fume Hood Work Surface, without Cup Sink Cutout, 72" Wide
Two year warranty

Key Features

  • Dished to retain spills,
  • Ideal for use with 72" wide bench fume hood

More About this Item

Choose from a wide range of accessories to enhance the performance of your fume hood.

These 1 1/4" thick blak epoxy resin work surfaces are required for all bench hoods. Sinks and cutouts sold separately.

For sale in U.S. only!

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat