ICI Quick Ship Cold Water Fitting for Fume Hood, without Vacuum Breaker
Representative image only.
ICI Quick Ship Cold Water Fitting for Fume Hood, without Vacuum Breaker
ICI Quick Ship | Mfr: CPL3185W-9RS| Item: UX-78981-01
Enhance your fume hood performance
$356.50USD / Each
$356.50USD / Each
Product Description
Key Features
  • Shipped loose and installed on site
  • 2 year warranty
More About this Item
Choose from a wide range of accessories to enhance the performance of your fume hood.
This remote control cold water fitting features a brass angle valve with a replaceable seat, an epoxy-coded brass nylon color-coded panel flange with goosenck without a vacuum breaker, a remote control rod assembly with plastic flange, and a tank nipple, locknut and washer.
For sale in U.S. only!
Two year warranty
Specifications
  • Compatible WithICI Quick Ship ducted fume hoods
  • DescriptionICI Quick Ship Cold Water Fitting for Fume Hood, without Vacuum Breaker