ICI Quick Ship Fittings for Fume Hood, Vacuum, Yellow
Representative image only.
ICI Quick Ship Fittings for Fume Hood, Vacuum, Yellow
ICI Quick Ship | Mfr: CPL3185N-LR-V| Item: UX-78980-98
Enhance your fume hood performance
$269.50USD / Each
$269.50USD / Each
Product Description
Key Features
  • May be used on either side of hood
  • Shipped loose and installed on site
  • 2 year warranty
More About this Item
Choose from a wide range of accessories to enhance the performance of your fume hood.
These remote control fittings feature a brass angle valve with a replaceable seat, an epoxy-coded panel flange with angle serrated hose connector, a remote control rod assembly with plastic flange, and a tank nipple, locknut and washer.
For sale in U.S. only!
Two year warranty
Specifications
  • Compatible WithICI Quick Ship ducted fume hoods
  • DescriptionICI Quick Ship Fittings for Fume Hood, Vacuum, Yellow