ICI Quick Ship Fume Hood Gravity Sash Stop
Representative image only.
ICI Quick Ship Fume Hood Gravity Sash Stop
ICI Quick Ship | Mfr: CPSP01| Item: UX-78981-13
Enhance your fume hood performance
$84.30USD / Each
$84.30USD / Each
Product Description
Key Features
  • Pivots out of the way when not needed
  • 2 year warranty
More About this Item
Choose from a wide range of accessories to enhance the performance of your fume hood.
The gravity sash stop can be added to any ICI Quick Ship fume hood as a safety and energy conservation measure. You can mount it at any distance above the lower deflactor vane to keep the sash from rising past that point.
Two year warranty
Specifications
  • Compatible WithICI Quick Ship ducted fume hoods
  • DescriptionICI Quick Ship Fume Hood Gravity Sash Stop
  • Warranty2 Years