ICI Quick Ship Transparent Dome Hood for Original Exhaust Snorkel
Representative image only.
ICI Quick Ship Transparent Dome Hood for Original Exhaust Snorkel
ICI Quick Ship | Mfr: CP70376982| Item: UX-78981-20
Safely captures contaminants
$420.50USD / Each
$420.50USD / Each
Product Description
Key Features
  • Attaches directly on mini hood, irrespective of arm size
  • Ergonomic handle design of both sides of hood make it easy to maneuver
  • 2 year warranty
More About this Item
Attached one of these optional exhaust hoods to your exhaust snorkel arm to capture contaminants.
Constructed of PET-G plastic, the transparent dome hood is designed to capture high dispersion contaminants.
For sale in U.S. only!
Two year warranty
Specifications
  • DescriptionTransparent Dome Hood for Original Exhaust Snorkel
  • Warranty2 Years