LabChem Calmagite Indicator for Hardness, 0.1% Aqueous; 125 mL

Representative image only.

LabChem Calmagite Indicator for Hardness, 0.1% Aqueous; 125 mL

LabChem | Mfr: LC128507| Item: UX-80014-51

Highest quality raw materials!

$31.30USD / Each

$31.30USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 2340 C
  • Purity Grade
  • Element/IonWater Hardness
  • Volume (mL)125
  • PackagingPoly Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionCalmagite Indicator, 0.1% Aqueous, for Hardness; 125 mL
Documents & Certificates
Reviews
Q&A