LabChem ORP Standard, 200 mV; 500 mL

Representative image only.

LabChem ORP Standard, 200 mV; 500 mL

LabChem | Mfr: LC180101| Item: UX-80029-98

Highest quality raw materials!

$107.75USD / Each

$107.75USD / Each

Buy It Together

Current Item

LabChem ORP Standard, 200 mV; 500 mL
LabChem ORP Standard, 200 mV; 500 mL
8002998

$107.75USD / Each
+ =
Total Price 238.80 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration200 mV
  • Purity Grade
  • ChemistryStandards
  • Element/IonORP
  • Volume (mL)500
  • PackagingAmber Poly Bottle
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionORP Standard, 200 mV; 500 mL
Reviews
Q&A