LabChem Sodium nitroprusside 0.05% w/v, 500 mL

Representative image only.

LabChem Sodium nitroprusside 0.05% w/v, 500 mL

LabChem | Mfr: LC247001| Item: UX-35205-45

Meet EPA method 351.1 and 351.2

$61.65USD / Each

$61.65USD / Each
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Specifications
  • DescriptionSodium nitroprusside 0.05% w/v, 500 mL
Reviews
Q&A