LabChem Water, Deionized, ACS Grade, ASTM Type I, 20 L

Representative image only.

LabChem Water, Deionized, ACS Grade, ASTM Type I, 20 L

LabChem | Mfr: LC267405| Item: UX-80055-01

Quality and consistency your lab can count on

$134.75USD / Each

$134.75USD / Each
Product Description

Key Features

  • Water meets ACS Reagent grade ASTM Type I standards
  • Water comes in 20 L size and packaged in a Cubitainer®

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaH2O
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight18.02
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, Deionized, ACS Grade, ASTM Type I; 20 L
Reviews
Q&A