Labconco FreeZone Freeze Drying Chamber, Clear, Tall; BT FD
Representative image only.
Labconco FreeZone Freeze Drying Chamber, Clear, Tall; BT FD
Labconco | Mfr: 7318700| Item: UX-03353-15
Easily attach flasks to your freeze dry system or contain samples in bulk
$3,397.50USD / Each
$3,775.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$3,397.50USD / Each
$3,775.00USD / Each
Buy It Together
Current Item
Labconco FreeZone Freeze Drying Chamber, Clear, Tall; BT FD
Labconco FreeZone Freeze Drying Chamber, Clear, Tall; BT FD
0335315
$3,397.50USD / Each
$3,775.00USD / Each
+ + +
Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD
0334831
$3,834.00USD / Each
$4,260.00USD / Each
+
Labconco FreeZone Stainless Steel Adapter, Straight; 3/4" to 3/4"
0335431
$45.18USD / Each
$50.20USD / Each
=
Total Price 8458.68 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Clear acrylic lid, gasket, vacuum release valve, and pass through for electrical cords.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Obtain visibility while freeze drying samples in bulk
  • Accommodates tall product shelves
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The tall acrylic drying chamber is ideal for freeze drying numerous samples in bulk. It features an 8” diameter bottom opening to the collector and fits a 10-1/8” or 14” diameter condenser.
1 Year Warranty
Specifications
  • Exterior Height (InchShort)24.625
  • DescriptionFreeze Drying Chamber, Clear, Tall; BT FD