Labconco LabSolutions™ Powder Detergent, Large Pail; 27.5 Lbs
Representative image only.
Labconco Mfr # 4422100 – Item # EW-98963-96
Removes wide range of lab contaminants
  • Rinses completely
  • Noncorrosive
  • Contains phosphates
  • Does approximately 400 loads
PRODUCT OPTIONS
$370.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)12.474
  • MaterialNonfoaming powder
  • Compatible WithGlassware washers
  • DescriptionPowder Detergent, Large Pail, 400 Loads; 27.5 Lbs
Includes
Scoop.
Key Features
  • Rinses completely
  • Noncorrosive
  • Contains phosphates
  • Does approximately 400 loads
More About this Item
LabSolutions detergents are specially formulated for Labconco glassware washers.
Nonfoaming powder detergent is specially formulated to remove laboratory contaminants including grease, agar, and protein digestives. Approximate pH of 1% solution is 12.5. Ideal for automatic glassware washers.