LaMotte 2999 Test Strips, Chlorine Dioxide, 0-10ppm, 50 Strips/Vial
Representative image only.
LaMotte 2999 Test Strips, Chlorine Dioxide, 0-10ppm, 50 Strips/Vial
LaMotte | Mfr: 2999| Item: UX-99532-33
Use these chemically treated paper strips as a quick test for sanitizer strength
$23.44USD / Each
$26.05USD / Each
$23.44USD / Each
$26.05USD / Each
Product Description
Key Features
  • Compare color change to included chart for easy-to-read results
  • 1 year warranty
More About this Item
Strips and color chart come supplied in a waterproof plastic vial.
Test strips permit an easy swirl and read way to determine chlorine dioxide levels from 0 to 10ppm.
1 Year Warranty
Specifications
  • TestChlorine dioxide
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionTest Strips, Chlorine Dioxide, 0-10ppm, 50 Strips/Vial