M-System W2MS-AA-R Potentiometer Transmitter; Dual-Output/4-20mA/100-240VAC

Representative image only.

M-System W2MS-AA-R Potentiometer Transmitter; Dual-Output/4-20mA/100-240VAC

M-System | Mfr: W2MS-AA-R/UL| Item: UX-68467-52

$339.57USD / Each
$399.50USD / Each

$339.57USD / Each
$399.50USD / Each
Product Description

Key Features

  • Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring
  • 2000 VAC Isolation
  • Standard DIN rail or surface mounting
  • Base socket included with modules
  • CE marking and UL/C-UL Class I Div 2 approval
  • 3 year warranty

More About this Item

M-System's Mini-M Series Signal Conditioners are designed to accept the largest range of process signal inputs and provide a standard and non-standard DC output.

The Mini-MW Series Signal Splitters provide a second isolated output of independent range, which give you the flexibility to add sophisticated distributed control or MIS monitoring to a local loop, without a worry about mismatched impedance, or the threat of one problem system's output signal impacting the other.

Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring.

3 Year Warranty
Specifications
  • Max Input ohms (ohms)10000
  • Min Input ohms (ohms)0
  • Max Output ch1 (mA)20
  • Min Output ch1 (mA)4
  • Power Supply (VDC)24
  • Max Operating Temperature (° C)55
  • Min Operating Temperature (° C)-5
  • Max Operating Temperature (° F)131
  • Min Operating Temperature (° F)23
  • Height (in)3 33/64
  • Width (in)1 5/32
  • Depth (in)4 7/8
  • Height (cm)8.9
  • Width (cm)2.9
  • Depth (cm)12.4
  • DescriptionPotentiometer Transmitter; Dual-Output/4-20mA/100-240VAC
  • Warranty3 Years
Reviews
Q&A