Mark-10 Coefficient of Friction (COF) Testing Module for IntelliMESUR® Software
Representative image only.
Mark-10 Coefficient of Friction (COF) Testing Module for IntelliMESUR® Software
Mark-10 | Mfr: IMF001| Item: UX-67801-08
Enhance your digital test frame with IntelliMESUR software with advanced feature modules
$560.00USD / Each
sign in for your price.
$560.00USD / Each
sign in for your price.
Product Description
Key Features
  • Field upgradeable—no need to ship it to the factory or call for service
  • Meets ASTM D1984 and other relevant standards
More About this Item
Characterize and analyze material behavior and measure static and dynamic coefficient of friction (COF) with optional software upgrades. Upgrades can be done at any time and easily upgraded in the field via activation code.
The coefficient of friction )COF) module enables the digital force tester to measure static and dynamic COF for various materials.
Specifications
  • Compatible WithTest frames with IntelliMESUR® software
  • DescriptionCoefficient of Friction (COF) Testing Module
  • Warranty3 years