Mettler Toledo 30705123 Cuvette Carousel XPathHolder™, 20/30mm
Representative image only.
Mettler Toledo 30705123 Cuvette Carousel XPathHolder™, 20/30mm
Mettler Toledo | Mfr: 30705123| Item: UX-83700-84
Optimize performance of your Mettler Toledo EasyPlus spectrophotometers
$47.50USD / Each
$47.50USD / Each
Product Description
Key Features
  • Expand analysis capabilities and make testing easier
  • Cuvette carousel with cuvette holder is suitable for use with 20 and 30 mm cuvettes
  • Only one carousel can be installed in the analysis compartment at a time
More About this Item
Choose from a wide range of accessories including cuvette carousels, replacement bulbs, and more for your Mettler Toledo EasyPlus spectrophotometers.
Cuvette holders provide a secure base for your sample, ensuring highly accurate measurement results.
Specifications
  • Pathlength (mm)20 and 30 mm
  • Materials of ConstructionPA66 Polyamide
  • DescriptionCuvette Carousel XPathHolder™, 20/30mm