Mettler Toledo 30705124 Cuvette Carousel XPathHolder™, 40/50mm/1"
Representative image only.
Mettler Toledo 30705124 Cuvette Carousel XPathHolder™, 40/50mm/1"
Mettler Toledo | Mfr: 30705124| Item: UX-83700-85
Optimize performance of your Mettler Toledo EasyPlus spectrophotometers
$47.50USD / Each
$47.50USD / Each
Product Description
Key Features
  • Expand analysis capabilities and make testing easier
  • Cuvette carousel with cuvette holder is suitable for use with 40 mm, 50 mm, and 1" cuvettes
  • Only one carousel can be installed in the analysis compartment at a time
More About this Item
Choose from a wide range of accessories including cuvette carousels, replacement bulbs, and more for your Mettler Toledo EasyPlus spectrophotometers.
Cuvette holders provide a secure base for your sample, ensuring highly accurate measurement results.
Specifications
  • Pathlength (mm)40 and 50 mm and 1 inch
  • Materials of ConstructionPA66 Polyamide
  • DescriptionCuvette Carousel XPathHolder™, 40/50mm/1"