Mettler Toledo EM40 EasyPlus Redox sensor
Representative image only.
Mettler Toledo EM40 EasyPlus Redox sensor
Mettler Toledo | Mfr: 30043106| Item: UX-94190-52
Enhance the versatility of your Mettler Toledo titrators
$258.00USD / Each
$258.00USD / Each
Buy It Together
Current Item
Mettler Toledo EM40 EasyPlus Redox sensor
Mettler Toledo EM40 EasyPlus Redox sensor
9419052
$258.00USD / Each
+ + =
Total Price 19283.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • ReDox with cable and BNC connector
  • Used with Easy OX and Easy Pro titrators
  • 1 year warranty
More About this Item
Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.
Replacement for sensor included in Easy OX kit.
1 Year Warranty
Specifications
  • Max mV (mV)2000
  • mV Resolution0.1
  • DescriptionEasyPlus Redox sensor