Mettler Toledo SmartCal StarterPac Moisture Analyzer Reference Substance, 12 Tests
Representative image only.
Mettler Toledo SmartCal StarterPac Moisture Analyzer Reference Substance, 12 Tests
Mettler Toledo | Mfr: 30005917| Item: UX-11336-34
Quick and Easy Performance Test for Moisture Analyzers
$695.00USD / Pkg of 12
$695.00USD / Pkg of 12
Product Description
Includes
12 test sachets, reporting templates, thermo-hygrometer, and validation documentation.
Key Features
  • Indicates incorrect performance immediately
  • Test temperatures include 70, 100, 130, 160°C (158, 212, 266, 320°F)
  • Includes product certificate
  • Offers a shelf life of 27 months
  • Three month warranty
More About this Item
SmartCal verifies the performance of your moisture analyzer in a 10-minute test, which is carried out in the same easy way as a regular measurement. Regular testing with SmartCal lets you provides documented evidence of the unit's performance over time. Incorrect performance is immediately indicated. The test instantly indicates adverse influences from the heating or weighing units. If the test is passed, you can trust your results.
This starter kit contains 12 test sachets plus all accessories you need including measurement protocols, reporting templates, a thermo/hygrometer, and validation documentation.
3 Month Warranty
Specifications
  • Accessory TypeFor use with Mettler Toledo Halogen Moisture Analyzer
  • DescriptionSmartCal StarterPac Moisture Analyzer Reference Substance, 12 Tests