Microflex DF850-M Vinyl Gloves, medium, 1000/cs
Representative image only.
Microflex DF850-M Vinyl Gloves, medium, 1000/cs
Microflex | Mfr: DF850-M| Item: UX-86232-26
Ideal for food processing or light industrial use
$133.60USD / Case of 1000
BUY MORE & SAVE
  • 1-2$133.60 USD / Case of 1000
  • 3+$120.24 USD / Case of 1000
$133.60USD / Case of 1000
BUY MORE & SAVE
  • 1-2$133.60 USD / Case of 1000
  • 3+$120.24 USD / Case of 1000
Product Description
Key Features
  • Tear resistance
  • Powder-free
More About this Item
Ambidextrous, 100% synthetic powder-free vinyl gloves alleviate the potential from adverse reactions from natural rubber proteins. Gloves are thicker for added tear resistance without sacrificing tactile sensitivity. Lightly beaded cuffs ensure hassle-free removal after use.
Specifications
  • Thickness (mil)6
  • MaterialVinyl
  • SizeMedium
  • Length (in)9
  • Length (cm)23
  • ColorClear
  • CuffBeaded
  • SterileNo
  • DescriptionVinyl Gloves, medium, 1000/cs