Myron L UltraPen PT3 UltraPen ORP and Temperature Pen

Representative image only.

Myron L UltraPen PT3 UltraPen ORP and Temperature Pen

Myron L | Mfr: PT3| Item: UX-05754-45

Waterproof, pocket-sized meters for water analysis testing

$355.45USD / Each

$355.45USD / Each

Buy It Together

Current Item

Myron L UltraPen PT3 UltraPen ORP and Temperature Pen
Myron L UltraPen PT3 UltraPen ORP and Temperature Pen
0575445

$355.45USD / Each
+ + +
Myron L PT1 ULTRAPEN Conductivity Pen
0575405

$294.55USD / Each
=
Total Price 3588.60 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

tester holster, lanyard, and 1.5V N alkaline battery.

Key Features

  • Up to 3 point calibration with 4, 7 and 10 buffer
  • Automatic temperature compensation
  • PTBT2 model with Bluetooh for data transfer

More About this Item

These waterproof pH and ORP testers offer anodized aircraft aluminum with protective coating and fully encapsulated electronics for durability. The LCD provides measurement reading with temperature available in both a live and hold measurement mode. Calibration features three point automatic buffer recognition for pH 4, 7, and 10 buffers.

Conformite EuropeenneIP 671 Year Warranty
Specifications
  • Meter TypeTester
  • Display typeLCD
  • mV Resolution1 mV
  • Temperature compensationAutomatic
  • WaterproofYes
  • Diameter (cm)1.5875
  • Length (in)6 3/4
  • Diameter (in)5/8
  • DescriptionUltraPen ORP and Temperature Pen
Reviews
Q&A