Novasina ClimMate SAL-SC 75 Sensor-Check Humidity Standard with Certificate
Representative image only.
Novasina ClimMate SAL-SC 75 Sensor-Check Humidity Standard with Certificate
Novasina | Mfr: 111 0859| Item: UX-37910-95
Keep your Novasina ClimMate Portable Water Activity Meter producing optimal results
$496.50USD / Each
$496.50USD / Each
Buy It Together
Current Item
Novasina ClimMate SAL-SC 75 Sensor-Check Humidity Standard with Certificate
Novasina ClimMate SAL-SC 75 Sensor-Check Humidity Standard with Certificate
3791095
$496.50USD / Each
+ =
Total Price 945.80 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
factory supplied certificate.
Key Features
  • Choose from a wide range of accessories including humidity standards, probes, and more
More About this Item
These replacement accessories are compatible with Novasina ClimMate Portable Water Activity meter, model 37910-38. Meter sold separately.
This humidity standard allows for field calibtration or adjustment of your ClimMate water activity meter. This standard is based on saturated salt solutions, housed in a plastic cylinder with a moisture-permeable membrane. This standard is compatible with the ClimMate water activity meter, model 37910-38, meter and probes sold separately.
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
Specifications
  • Compatible WithClimMate Portable Water Activity (aW) Meter 37910-38
  • DescriptionClimMate SAL-SC 75 Sensor-Check Humidity Standard with Certificate