Fytertech Spilfyter Grab & Go Hazmat Bucket Spill Kit, Absorbs 4 gallons
Representative image only.
Fytertech Spilfyter Grab & Go Hazmat Bucket Spill Kit, Absorbs 4 gallons
Fytertech | Mfr: 205304| Item: UX-86424-14
Designed for portability and quick response to small and medium spills
$148.25USD / Each
$148.25USD / Each
Product Description
Includes
Kit includes four 3" x 4' socks, three 16" x 18" pads, one pair of nitrile gloves, one pair of goggles, and one 5-gallon plastic bucket with snap-on lid.
Key Features
  • Snap-on lid protects contents
More About this Item
Hazmat sorbents are specifically designed to absorb aggressive fluids such as acids and bases. Ideal for laboratory use.
Specifications
  • Absorption4 gal. (16 L)
  • DescriptionHazmat Bucket Spill Kit, Absorbs 4 gallons