Cole-Parmer Soil pH Testing Kit; Meter, Probe, Beakers, and Buffers
Representative image only.
Cole-Parmer Soil pH Testing Kit; Meter, Probe, Beakers, and Buffers
Cole-Parmer | Item: EW-25000-43
Convenient kit provides the instruments and equipment you need for testing the pH of soil
$839.50USD / Each
$839.50USD / Each
Buy It Together
Current Item
Cole-Parmer Soil pH Testing Kit; Meter, Probe, Beakers, and Buffers
Cole-Parmer Soil pH Testing Kit; Meter, Probe, Beakers, and Buffers
2500043
$839.50USD / Each
+
Environmental Express Electrode pH Soil Glass BNC
0599262
$232.02USD / Each
$257.80USD / Each
=
Total Price 1071.52 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
pH meter with ATC probe, protective rubber boot, and batteries; soil pH electrode; five glass beakers (50, 100, 250, 600, 1000 mL); and five each of pH 4.01, 7.00, and 10.00 buffers and rinse solution pouches.
Key Features
  • Obtain soil measurements without the "suspension effect"
More About this Item
The rugged Oakton® pH 6+ handheld pH meter features high, ±0.01 pH unit accuracy with up to five-point push-button calibration and autobuffer recognition. Switch between pH and temperature measurements with the press of a button. Automatic temperature compensation (ATC) ensures precise measurements from 0 to 100°C. Protective rubber boot that also acts as a meter stand.
The refillable, soil pH electrode has an extra-wide 7.5 mm ceramic junction to prevent clogging and provide a fast response. Soil electrode has a maximum use temperature of 176°F (80°C), measures 145 mm L x 12 mm diameter, and uses a silver/silver-chloride (Ag/AgCl) reference cell.
The kit is rounded off with Pyrex® glass beakers and Oakton® pH calibration buffer pouches. The graduated beakers feature pouring spouts and uniform wall thickness for optimum balance between thermal shock resistance and mechanical strength. All five beaker sizes have an extra large marking spot — perfect for sample labeling. An assortment of pH 4.01, 7.00, and 10.00 buffers and rinse solution help keep your testing system running smoothly and accurately. Don't do more dishes, calibrate the probe by placing it directly in the pouch.
Specifications
  • Meter TypePortable
  • Display typeLCD
  • ConnectionsBNC
  • CalibrationUp to 5 points
  • Max Temperature (° C)80
  • Max Temperature (° F)176
  • Temperature Resolution0.1
  • Temperature compensationAutomatic
  • Battery life (hr)700
  • DescriptionSoil pH Testing Kit; Meter, Probe, Beakers, and Buffers