Pomona Electronics 5953 Retractable/Adjustable Needle Tip Test Probe Set
Representative image only.
Pomona Electronics 5953 Retractable/Adjustable Needle Tip Test Probe Set
Pomona Electronics | Mfr: 5953A| Item: UX-20009-36
Save time and money with versatile kits
$40.50USD / Each
$45.00USD / Each
$40.50USD / Each
$45.00USD / Each
Product Description
Key Features
  • Retractable piercing/needle-tip probe is fully insulated to within 1/8” of the tip
  • Adjustable tip from 0 to 3” (0 to 76mm) length
  • Banana plug is designed to fit most popular DMM jacks
  • 4 ft silicone insulated test lead wire for high temperature resistance and flexibility
  • 3 month warranty
More About this Item
Enhance the versitility of your mulitmeter with these Pomona Test Leads and Kits.
3 Month Warranty
Specifications
  • DescriptionRetractable/Adjustable Needle Tip Test Probe Set