Pomona Electronics B-12 Multi-Stacking Banana Plug Patch Cord, 12" (black)

Representative image only.

Pomona Electronics B-12 Multi-Stacking Banana Plug Patch Cord, 12" (black)

Pomona Electronics | Mfr: B-12-0| Item: UX-20009-72

Available for sale only in the United States and Canada

$7.82USD / Each
$8.69USD / Each

$7.82USD / Each
$8.69USD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Multi-Stacking Banana Plug Patch Cord

1 Year Warranty
Specifications
  • Length (in)12
  • Length (cm)30.5
  • DescriptionMulti-Stacking Banana Plug Patch Cord, 12" (black)
  • Warranty1 year
Reviews
Q&A