Pomona Electronics B-48 Multi-Stacking Banana Plug Patch Cord, 48" (Red)

Representative image only.

Pomona Electronics B-48 Multi-Stacking Banana Plug Patch Cord, 48" (Red)

Pomona Electronics | Mfr: B-48-2| Item: UX-20009-81

Available for sale only in the United States and Canada

$8.18USD / Each
$9.09USD / Each

$8.18USD / Each
$9.09USD / Each
Product Description

Key Features

  • 3 month warranty

More About this Item

Multi-Stacking Banana Plug Patch Cord

3 Month Warranty
Specifications
  • Length (in)48
  • Length (cm)122
  • DescriptionMulti-Stacking Banana Plug Patch Cord, 48" (Red)
  • Warranty3 month
Reviews
Q&A