Pomona Electronics Solar MC4 Clamp Test Lead Set
Representative image only.
Pomona Electronics Solar MC4 Clamp Test Lead Set
Pomona Electronics | Mfr: PVLEAD3| Item: UX-20051-07
Connect to test tools or clamp meters with these photovoltaic test leads
$96.30USD / Each
$107.00USD / Each
$96.30USD / Each
$107.00USD / Each
Product Description
Includes
Set of black and red MC4 solar power clamp test leads.
Key Features
  • Ensures safe measurements on photovoltaic (PV) modules and systems
  • Connect to instruments that accept 4 mm sheathed banana plugs
  • Nickel plated contacts
  • 1 year warranty
More About this Item
Monitor solar photovoltaic (PV) systems in solar power plants with these test lead sets. Users can connect to test tools or clamp meters with two different options.
Creates connection between the solar panel and inverter for measuring with a clamp meter. The black test lead is an MC4 (male) breakout to 12-inch MC4 (female) and 36-inch 4 mm sheathed banana plug test lead. The red test lead is an MC4 (female) breakout to 12-inch MC4 (male) and 36-inch 4 mm sheathed banana plug test lead.
1 Year Warranty
Specifications
  • DescriptionSolar MC4 Clamp Test Lead Set