Pomona Electronics Static Free Floor Mat, Conductive with Grounding Kit, 3' x 4'
Representative image only.
Pomona Electronics Static Free Floor Mat, Conductive with Grounding Kit, 3' x 4'
Pomona Electronics | Mfr: SCMAT3| Item: UX-20051-10
Devices to monitor operator's path-to-ground both continuously and as needed
$215.10USD / Each
$239.00USD / Each
$215.10USD / Each
$239.00USD / Each
Product Description
Key Features
  • Quickly test all personal grounding devices with the combo wrist strap and footwear tester
  • Continuously monitor a worksurface path-to-ground of less than 10 MΩ with the multi-mount continuous monitor
  • Protect operator's with static free floor mats and foot straps
  • Provides a flooring surface that does not generate a static charge and will act as part of the path to ground for an operator wearing ESD footwear
  • The resistance range of the surface layer remains low enough to act as a path to ground with the normal build up of dirt and grime on the mat
  • Lays flat, where edges will not curl
  • Designed for use in high traffic areas and good for use under chairs and carts
  • Works with ESD floor tiles and ESD floor finishes
  • Use to protect and minimize maintenance of ESD flooring tiles and finish
  • Complies to ANSI/ESD S20.20
  • 1 year warranty
More About this Item
Working with static sensitive devices? Pomona enables you to work with confidence. Multiple options allow you to build out a kit that works for you and protects your devices from damage due to static electricity.
1 Year Warranty
Specifications
  • DescriptionStatic Free Floor Mat, Conductive with Grounding Kit, 3' x 4'