Qsonica 4417 Ultrasonic Cell Distruptor Microtip Probe, 1/16" dia, 0.2 - 5 mL
Representative image only.
Qsonica 4417 Ultrasonic Cell Distruptor Microtip Probe, 1/16" dia, 0.2 - 5 mL
Qsonica | Mfr: 4417| Item: UX-04712-61
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
$380.00USD / Each
$380.00USD / Each
Buy It Together
Current Item
Qsonica 4417 Ultrasonic Cell Distruptor Microtip Probe, 1/16" dia, 0.2 - 5 mL
Qsonica 4417 Ultrasonic Cell Distruptor Microtip Probe, 1/16" dia, 0.2 - 5 mL
0471261
$380.00USD / Each
+ + =
Total Price 8356.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.