Reagents Acetone, Purified, Amber Glass Bottle, 1 L

Representative image only.

Reagents Acetone, Purified, Amber Glass Bottle, 1 L

Reagents | Mfr: C2102500-1C| Item: UX-78914-84

Manufactured using the highest quality raw materials in ISO 9001 facility

$81.10USD / Each

$81.10USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.8
  • Solubility information: miscible
  • 1 L amber glass bottle
  • 20 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Purity GradePurified
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Propanone, Dimethyl ketone
  • Volume (mL)1000
  • Formula Weight58.08
  • Vapor Pressure (torr)174.764
  • PackagingAmber glass bottle
  • DOT InformationACETONE, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1090
  • CAS Number67-64-1
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, Purified, Amber Glass Bottle, 1 L
  • Warranty20 years
Documents & Certificates
Reviews
Q&A