Reagents Acetone, Purified, Blue Winpak® Container, 20 L

Representative image only.

Reagents Acetone, Purified, Blue Winpak® Container, 20 L

Reagents | Mfr: C2102500-20E7| Item: UX-78926-77

Manufactured using the highest quality raw materials in ISO 9001 facility

$614.00USD / Each

$614.00USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.8
  • Solubility information: miscible
  • 20 L blue Winpak container
  • 20 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Purity GradePurified
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • Synonyms2-Propanone, Dimethyl ketone
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight58.08
  • Vapor Pressure (torr)174.764
  • PackagingWinpak container
  • DOT InformationACETONE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-64-1
  • UN NumberUN1090
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, Purified, Blue Winpak® Container, 20 L
  • Warranty20 years
Documents & Certificates
Reviews
Q&A