Reagents Ammonium Chloride, Purified, Natural Poly Container, 3 kg

Representative image only.

Reagents Ammonium Chloride, Purified, Natural Poly Container, 3 kg

Reagents | Mfr: C2132000-3A5| Item: UX-62283-88

Manufactured using the highest quality raw materials in ISO 9001 facility

$87.30USD / Each

$87.30USD / Each
Product Description

Key Features

  • White solid
  • Density: 1.53
  • Melting point: 337.8 °C (640.04 °F) with decomposition
  • Solubility information: 395 g/L at 25 °C (77 °F)
  • pH range: 4.5 to 5.5
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNH4Cl
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • Formula Weight53.49
  • Vapor Pressure (torr)0.9976
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number12125-02-9
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)520
  • Boiling Point F (° F)968
  • DescriptionAmmonium Chloride, Purified, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A