Reagents Ammonium Chloride, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Ammonium Chloride, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2132000-500A5| Item: UX-78910-24

Manufactured using the highest quality raw materials in ISO 9001 facility

$39.80USD / Each

$39.80USD / Each
Product Description

Key Features

  • White solid
  • Density: 1.53
  • Melting point: 337.8 °C (640.04 °F) with decomposition
  • Solubility information: 395 g/L at 25 °C (77 °F)
  • pH range: 4.5 to 5.5
  • 500g wide-mouth container
  • 20 years warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNH4Cl
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight53.49
  • Vapor Pressure (torr)0.9976
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number12125-02-9
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)520
  • Boiling Point F (° F)968
  • DescriptionAmmonium Chloride, Purified, Wide-Mouth Container, 500 g
  • Warranty20 years
Documents & Certificates
Reviews
Q&A